Tisk…

1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one 98%

BLDPBD00970964-25GEA 5560 CZK
BLDPBD00970964-25G BLDPBD00970964-1G BLDPBD00970964-5G
1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one 98%
1-Cyclopropyl-2-(triphenyl-lambda5-phosphanylidenE)ethan-1-one

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₂₃H₂₁OP
MW: 344,3860 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22124430
CAS číslo: 7691-76-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR