Tisk…

3-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)propanoic acid 97%

BLDPBD00971561-250EA 3570 CZK
BLDPBD00971561-250 BLDPBD00971561-1G BLDPBD00971561-100
3-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)propanoic acid 97%
3-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₂H₁₄O₄
MW: 222,2372 g/mol
Storage Temperature: Refrigerator
CAS číslo: 1220710-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR