Tisk…

Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate 95%

BLDPBD00972075-250EA 1320 CZK
BLDPBD00972075-250 BLDPBD00972075-1G
Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate 95%
Ethyl (2E)-2-cyano-3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₈H₂₂BNO₄
MW: 327,1826 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20527090
CAS číslo: 2828447-15-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR