Tisk…

Roflumilast N-oxide 98%

BLDPBD00972179-250EA 15080 CZK
BLDPBD00972179-250 BLDPBD00972179-2MG BLDPBD00972179-25M BLDPBD00972179-100 BLDPBD00972179-5MG BLDPBD00972179-10M BLDPBD00972179-50M
Roflumilast N-oxide 98%
Roflumilast N-oxide

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₁₇H₁₄Cl₂F₂N₂O₄
MW: 419,21 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04112984
CAS číslo: 292135-78-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR