Tisk…

1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one 95%

BLDPBD00973555-250EA 5280 CZK
BLDPBD00973555-250 BLDPBD00973555-1G BLDPBD00973555-100
1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one 95%
1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₇BrF₂O₂
MW: 265,0515 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188449
CAS číslo: 1215994-75-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR