Tisk…

Diethyl [2,2'-bipyridine]-6,6'-dicarboxylate 97%

BLDPBD00973645-5GEA 3250 CZK
BLDPBD00973645-5G BLDPBD00973645-250 BLDPBD00973645-25G BLDPBD00973645-1G
Diethyl [2,2'-bipyridine]-6,6'-dicarboxylate 97%
Diethyl [2,2'-bipyridine]-6,6'-dicarboxylate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₆H₁₆N₂O₄
MW: 300,3092 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11042477
CAS číslo: 65739-40-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR