Tisk…

4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%

BLDPBD00974239-250EA 9500 CZK
BLDPBD00974239-250 BLDPBD00974239-1G BLDPBD00974239-100
4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione 95%
4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₁H₁₁NO₂
MW: 189,2105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00666096
CAS číslo: 5488-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR