Tisk…

Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98%

BLDPBD00975439-1GEA 18270 CZK
BLDPBD00975439-1G BLDPBD00975439-100 BLDPBD00975439-250
Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 98%
Tetrahydro-2H-pyran-4-yl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₉H₂₇BN₂O₅
MW: 374,2391 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33022343
CAS číslo: 2828431-82-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR