Tisk…

Amino-PEG5-t-butyl ester 98%

BLDPBD00976292-1GEA 7100 CZK
BLDPBD00976292-1G BLDPBD00976292-100 BLDPBD00976292-250
Amino-PEG5-t-butyl ester 98%
Amino-PEG5-t-butyl ester

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₇H₃₅NO₇
MW: 365,462 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25424125
CAS číslo: 1446282-18-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR