Tisk…

2-(1-Carbamoylpiperidin-4-yl)acetic acid 95%

BLDPBD00976312-250EA 4370 CZK
BLDPBD00976312-250 BLDPBD00976312-1G BLDPBD00976312-100
2-(1-Carbamoylpiperidin-4-yl)acetic acid 95%
2-(1-Carbamoylpiperidin-4-yl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₁₄N₂O₃
MW: 186,2084 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20638954
CAS číslo: 279236-52-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR