Tisk…

4-Bromo-6-fluoroisoquinoline 95%

BLDPBD00977850-100EA 7700 CZK
BLDPBD00977850-100 BLDPBD00977850-250
4-Bromo-6-fluoroisoquinoline 95%
4-Bromo-6-fluoroisoquinoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₅BrFN
MW: 226,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28661296
CAS číslo: 1416500-78-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR