Tisk…

6-(Trifluoromethoxy)quinolin-3-amine 95%

BLDPBD00984287-100EA 5160 CZK
BLDPBD00984287-100 BLDPBD00984287-1G BLDPBD00984287-250
6-(Trifluoromethoxy)quinolin-3-amine 95%
6-(Trifluoromethoxy)quinolin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₇F₃N₂O
MW: 228,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27941211
CAS číslo: 1051372-65-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR