Tisk…

5-Chloro-3-ethenyl-2-fluoropyridine 95% stabilizovaný


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

BLDPBD00984384-100EA 5620 CZK
BLDPBD00984384-100 BLDPBD00984384-250
5-Chloro-3-ethenyl-2-fluoropyridine 95% stabilizovaný
5-Chloro-3-ethenyl-2-fluoropyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₅ClFN
MW: 157,5700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22191412
CAS číslo: 1824297-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR