Tisk…

2-Fluoro-4-methylpentanoic acid 95%

BLDPBD00985815-100EA 4450 CZK
BLDPBD00985815-100 BLDPBD00985815-1G BLDPBD00985815-250
2-Fluoro-4-methylpentanoic acid 95%
2-Fluoro-4-methylpentanoic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₁₁FO₂
MW: 134,1488 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24089385
CAS číslo: 6087-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR