Tisk…

1-Benzofuran-6-amine 95%

BLDPBD00988324-5GEA 28280 CZK
BLDPBD00988324-5G BLDPBD00988324-100 BLDPBD00988324-1G BLDPBD00988324-250
1-Benzofuran-6-amine 95%
1-Benzofuran-6-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₈H₇NO
MW: 133,1473 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110252
CAS číslo: 110677-54-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR