Tisk…

Methyl 2-(chlorosulphonyl)-5-methylbenzoate 95%

BLDPBD00989909-1GEA 9810 CZK
BLDPBD00989909-1G BLDPBD00989909-250 BLDPBD00989909-100
Methyl 2-(chlorosulphonyl)-5-methylbenzoate 95%
Methyl 2-(chlorosulphonyl)-5-methylbenzoate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₉ClO₄S
MW: 248,6834 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09746476
CAS číslo: 1153230-09-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR