Tisk…

Oxepane-4-carboxylic acid 98%

BLDPBD00990947-250EA 6930 CZK
BLDPBD00990947-250 BLDPBD00990947-100 BLDPBD00990947-1G
Oxepane-4-carboxylic acid 98%
Oxepane-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₁₂O₃
MW: 144,1684 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21124931
CAS číslo: 933747-23-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR