Tisk…

3-(2-Bromo-6-methylphenyl)propanoic acid 95%

BLDPBD00992042-250EA 10010 CZK
BLDPBD00992042-250 BLDPBD00992042-50M BLDPBD00992042-1G BLDPBD00992042-100
3-(2-Bromo-6-methylphenyl)propanoic acid 95%
3-(2-Bromo-6-methylphenyl)propanoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁BrO₂
MW: 243,1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392247
CAS číslo: 1261861-12-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR