Tisk…

1-(4-Fluoro-3-methylphenyl)ethan-1-amine 95%

BLDPBD00993489-50MEA 3420 CZK
BLDPBD00993489-50M BLDPBD00993489-100 BLDPBD00993489-1G BLDPBD00993489-250
1-(4-Fluoro-3-methylphenyl)ethan-1-amine 95%
1-(4-Fluoro-3-methylphenyl)ethan-1-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₁₂FN
MW: 153,1967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12805315
CAS číslo: 105321-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR