Tisk…

2-Aminobenzene-1-sulphonyl fluoride 95%

BLDPBD00994601-250EA 2710 CZK
BLDPBD00994601-250 BLDPBD00994601-100 BLDPBD00994601-1G
2-Aminobenzene-1-sulphonyl fluoride 95%
2-Aminobenzene-1-sulphonyl fluoride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₆FNO₂S
MW: 175,1807 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007704
CAS číslo: 392-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR