Tisk…

3-Amino-4-hydroxy-N-methylbenzamide 95%

BLDPBD00997382-50MEA 3030 CZK
BLDPBD00997382-50M BLDPBD00997382-100 BLDPBD00997382-1G BLDPBD00997382-250
3-Amino-4-hydroxy-N-methylbenzamide 95%
3-Amino-4-hydroxy-N-methylbenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₁₀N₂O₂
MW: 166,1772 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998311
CAS číslo: 1243286-96-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR