Tisk…

Ethyl 4-(hydroxymethyl)-1H-pyrazole-3-carboxylate 95%

BLDPBD01000222-1GEA 24460 CZK
BLDPBD01000222-1G BLDPBD01000222-100 BLDPBD01000222-250
Ethyl 4-(hydroxymethyl)-1H-pyrazole-3-carboxylate 95%
Ethyl 4-(hydroxymethyl)-1H-pyrazole-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₁₀N₂O₃
MW: 170,1659 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16620141
CAS číslo: 61453-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR