Tisk…

(E)-3-(2-Methylfuran-3-yl)acrylic acid 95%

BLDPBD01001142-1GEA 38310 CZK
BLDPBD01001142-1G BLDPBD01001142-250 BLDPBD01001142-100 BLDPBD01001142-50M
(E)-3-(2-Methylfuran-3-yl)acrylic acid 95%
(E)-3-(2-Methylfuran-3-yl)acrylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₈O₃
MW: 152,1473 g/mol
Storage Temperature: Refrigerator
CAS číslo: 79190-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR