Tisk…

Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate 95%

BLDPBD01003996-1GEA 27360 CZK
BLDPBD01003996-1G BLDPBD01003996-100 BLDPBD01003996-50M BLDPBD01003996-250
Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate 95%
Ethyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₈BrN₃O₂
MW: 270,0827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23708550
CAS číslo: 1427376-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR