Tisk…

6-Chloro-1,5-naphthyridin-2(1H)-one 95%

BLDPBD01004934-250EA 9300 CZK
BLDPBD01004934-250 BLDPBD01004934-50M BLDPBD01004934-100
6-Chloro-1,5-naphthyridin-2(1H)-one 95%
6-Chloro-1,5-naphthyridin-2(1H)-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₅ClN₂O
MW: 180,5911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543817
CAS číslo: 1394003-98-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR