Tisk…

1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride 95%

BLDPBD01005405-5GEA 27680 CZK
BLDPBD01005405-5G BLDPBD01005405-250 BLDPBD01005405-1G
1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride 95%
1H,2H,3H,4H,5H-Pyrido[4,3-b]indole-8-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₂H₁₃ClN₂O₂
MW: 252,6968 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971425
CAS číslo: 1156899-12-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR