Tisk…

(4S)-5,8-Difluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride 95%

BLDPBD01010699-100EA 8580 CZK
BLDPBD01010699-100 BLDPBD01010699-250
(4S)-5,8-Difluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride 95%
(4S)-5,8-Difluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₀ClF₂NO
MW: 221,6316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24435651
CAS číslo: 1807940-80-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR