Tisk…

8-Fluoroisoquinoline-5-carbonitrile 95%

BLDPBD01013550-100EA 2230 CZK
BLDPBD01013550-100 BLDPBD01013550-1G BLDPBD01013550-250
8-Fluoroisoquinoline-5-carbonitrile 95%
8-Fluoroisoquinoline-5-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₅FN₂
MW: 172,1585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26407892
CAS číslo: 1368344-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR