Tisk…

5-Chloropentan-2-ol 95%

BLDPBD01014289-250EA 3420 CZK
BLDPBD01014289-250
5-Chloropentan-2-ol 95%
5-Chloropentan-2-ol

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₅H₁₁ClO
MW: 122,5932 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19231978
CAS číslo: 15146-94-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR