Tisk…

2-Amino-2-(4-chloro-3-fluorophenyl)ethan-1-ol hydrochloride 95%

BLDPBD01018138-100EA 4310 CZK
BLDPBD01018138-100 BLDPBD01018138-250
2-Amino-2-(4-chloro-3-fluorophenyl)ethan-1-ol hydrochloride 95%
2-Amino-2-(4-chloro-3-fluorophenyl)ethan-1-ol hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₁₀Cl₂FNO
MW: 226,0755 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23144294
CAS číslo: 1427380-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR