Tisk…

2-Formyl-N,N-dimethyl-1H-imidazole-1-sulphonamide 95%

BLDPBD01021296-250EA 6730 CZK
BLDPBD01021296-250 BLDPBD01021296-100 BLDPBD01021296-1G BLDPBD01021296-50M
2-Formyl-N,N-dimethyl-1H-imidazole-1-sulphonamide 95%
2-Formyl-N,N-dimethyl-1H-imidazole-1-sulphonamide

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₉N₃O₃S
MW: 203,2190 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD25959869
CAS číslo: 167704-98-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR