Tisk…

tert-Butyl 2-carbamothioyl-2,2-dimethylacetate 95%

BLDPBD01021592-100EA 4020 CZK
BLDPBD01021592-100 BLDPBD01021592-250 BLDPBD01021592-1G
tert-Butyl 2-carbamothioyl-2,2-dimethylacetate 95%
tert-Butyl 2-carbamothioyl-2,2-dimethylacetate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₁₇NO₂S
MW: 203,3018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24625932
CAS číslo: 75235-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR