Tisk…

N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide 95%

BLDPBD01023287-100EA 2090 CZK
BLDPBD01023287-100 BLDPBD01023287-5G BLDPBD01023287-250 BLDPBD01023287-1G
N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide 95%
N-(2-(1H-Indol-3-yl)ethyl)-2-aminoacetamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₂H₁₅N₃O
MW: 217,2670 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08282638
CAS číslo: 122902-82-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR