Tisk…

Methyl 6-amino-2-bromo-3-methoxybenzoate 95%

BLDPBD01027941-50MEA 3620 CZK
BLDPBD01027941-50M BLDPBD01027941-100 BLDPBD01027941-250 BLDPBD01027941-1G
Methyl 6-amino-2-bromo-3-methoxybenzoate 95%
Methyl 6-amino-2-bromo-3-methoxybenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₁₀BrNO₃
MW: 260,0846 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19629015
CAS číslo: 1340366-76-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR