Tisk…

3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-amine 95%

BLDPBD01029333-100EA 5480 CZK
BLDPBD01029333-100 BLDPBD01029333-50M BLDPBD01029333-1G BLDPBD01029333-250
3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-amine 95%
3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₅H₇F₂N₃
MW: 147,1260 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28963069
CAS číslo: 1801762-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR