Tisk…

Ethyl[(3-methylphenyl)methyl]amine 95%

BLDPBD01030487-250EA 1630 CZK
BLDPBD01030487-250 BLDPBD01030487-100 BLDPBD01030487-1G BLDPBD01030487-5G
Ethyl[(3-methylphenyl)methyl]amine 95%
Ethyl[(3-methylphenyl)methyl]amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₅N
MW: 149,2328 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048518
CAS číslo: 209051-77-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR