Tisk…

4-Phenoxypyrimidin-2-amine 95%

BLDPBD01031164-1GEA 19840 CZK
BLDPBD01031164-1G BLDPBD01031164-100 BLDPBD01031164-5G BLDPBD01031164-250
4-Phenoxypyrimidin-2-amine 95%
4-Phenoxypyrimidin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₉N₃O
MW: 187,1980 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463193
CAS číslo: 882767-05-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR