Tisk…

5-Acetylfuran-2-carboxylic acid 95%

BLDPBD01031606-1GEA 22350 CZK
BLDPBD01031606-1G BLDPBD01031606-250 BLDPBD01031606-100 BLDPBD01031606-50M
5-Acetylfuran-2-carboxylic acid 95%
5-Acetylfuran-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₆O₄
MW: 154,1201 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18431708
CAS číslo: 13341-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR