Tisk…

(5-Cyclopropylfuran-2-yl)methanol 95%

BLDPBD01031618-1GEA 14340 CZK
BLDPBD01031618-1G BLDPBD01031618-100 BLDPBD01031618-250
(5-Cyclopropylfuran-2-yl)methanol 95%
(5-Cyclopropylfuran-2-yl)methanol

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₈H₁₀O₂
MW: 138,1638 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28970751
CAS číslo: 1935403-22-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR