Tisk…

5-Methyl-1-(piperidin-4-yl)-1H-pyrazole-4-carboxylic acid 95%

BLDPBD01033631-250EA 6590 CZK
BLDPBD01033631-250 BLDPBD01033631-1G BLDPBD01033631-100
5-Methyl-1-(piperidin-4-yl)-1H-pyrazole-4-carboxylic acid 95%
5-Methyl-1-(piperidin-4-yl)-1H-pyrazole-4-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₁₅N₃O₂
MW: 209,2450 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28397372
CAS číslo: 1780811-54-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR