Tisk…

3-Amino-4-bromobenzene-1-sulphonamide 95%

BLDPBD01034981-250EA 2540 CZK
BLDPBD01034981-250 BLDPBD01034981-1G BLDPBD01034981-100
3-Amino-4-bromobenzene-1-sulphonamide 95%
3-Amino-4-bromobenzene-1-sulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₇BrN₂O₂S
MW: 251,1010 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09949821
CAS číslo: 100367-90-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR