Tisk…

Methyl 2-methoxy-3-methylphenylacetate 95%

BLDPBD01037141-1GEA 2230 CZK
BLDPBD01037141-1G BLDPBD01037141-250
Methyl 2-methoxy-3-methylphenylacetate 95%
Methyl 2-methoxy-3-methylphenylacetate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₄O₃
MW: 194,2271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399542
CAS číslo: 1261827-91-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR