Tisk…

3-Methyl-1-(naphthalen-1-yl)butan-1-amine hydrochloride 95%

BLDPBD01042890-100EA 3880 CZK
BLDPBD01042890-100 BLDPBD01042890-250
3-Methyl-1-(naphthalen-1-yl)butan-1-amine hydrochloride 95%
3-Methyl-1-(naphthalen-1-yl)butan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₅H₂₀ClN
MW: 249,7790 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28126515
CAS číslo: 1204595-16-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR