Tisk…

4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline 95%

BLDPBD01044192-1GEA 3910 CZK
BLDPBD01044192-1G BLDPBD01044192-100 BLDPBD01044192-5G BLDPBD01044192-250
4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline 95%
4-Fluoro-N-(furan-2-ylmethyl)-2-methylaniline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₂FNO
MW: 205,2282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01135302
CAS číslo: 418785-68-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR