Tisk…

3-sulphopropanoic acid 95%

BLDPBD01044758-100EA 6470 CZK
BLDPBD01044758-100 BLDPBD01044758-250 BLDPBD01044758-50M
3-sulphopropanoic acid 95%
3-sulphopropanoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₃H₆O₅S
MW: 154,1417 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19347139
CAS číslo: 44826-45-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR