Tisk…

1-(Cyclohexylamino)-2-methylpropan-2-ol 98%

BLDPBD01044809-1GEA 15310 CZK
BLDPBD01044809-1G BLDPBD01044809-250 BLDPBD01044809-100
1-(Cyclohexylamino)-2-methylpropan-2-ol 98%
1-(Cyclohexylamino)-2-methylpropan-2-ol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₁₀H₂₁NO
MW: 171,2798 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11916062
CAS číslo: 7527-65-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR