Tisk…

Methyl 3-bromo-5-(cyanomethyl)benzoate 95%

BLDPBD01048006-250EA 3940 CZK
BLDPBD01048006-250 BLDPBD01048006-100 BLDPBD01048006-1G
Methyl 3-bromo-5-(cyanomethyl)benzoate 95%
Methyl 3-bromo-5-(cyanomethyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₈BrNO₂
MW: 254,0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18378610
CAS číslo: 878741-84-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR