Tisk…

5,6-Dimethoxyindole 98%

BLDPBD0105-5GEA 6560 CZK
BLDPBD0105-5G BLDPBD0105-250MG BLDPBD0105-10G BLDPBD0105-1G BLDPBD0105-100MG BLDPBD0105-25G
5,6-Dimethoxyindole 98%
5,6-Dimethoxyindole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005675
CAS číslo: 14430-23-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR