Tisk…

2-(Dimethylamino)pyridine-3-carboxylic acid hydrochloride 95%

BLDPBD01056252-100EA 2170 CZK
BLDPBD01056252-100 BLDPBD01056252-250
2-(Dimethylamino)pyridine-3-carboxylic acid hydrochloride 95%
2-(Dimethylamino)pyridine-3-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₁ClN₂O₂
MW: 202,6381 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505431
CAS číslo: 1177289-23-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR