Tisk…

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%

BLDPBD0106-5GEA 257 CZK
BLDPBD0106-5G BLDPBD0106-500G BLDPBD0106-100G BLDPBD0106-25G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid 98%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₁NO₂
MW: 177,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144533
CAS číslo: 74163-81-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR